missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monostearate, Powder, Food Grade, Spectrum™ Chemical

Catalog No. 18603000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603000 500 g Poly Bottle
18603001 2.5 kg Poly Pail
18603002 12 kg Poly Pail
18603003 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603000 Supplier Spectrum Chemical Mfg Cor Supplier No. GL149500GM
Only null left

GL149, 31566-31-1, C21H42O4

Spectrum™ Chemical Glyceryl Monostearate, Powder, Food Grade is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color nor odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. In addition to use as an emulsifier, it is also an anti-caking and thickening agent.

Chemical Identifiers

CAS 31566-31-1
Molecular Formula C21H42O4
Molecular Weight (g/mol) 358.56
InChI Key VBICKXHEKHSIBG-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxypropyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Grade Food Grade