missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monostearate, Powder, Food Grade, Spectrum™ Chemical

GL149, 31566-31-1, C21H42O4

Supplier:  Spectrum Chemical Mfg Cor GL14912KG

 View more versions of this product

Catalog No. 18603002


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Glyceryl Monostearate, Powder, Food Grade is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color nor odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. In addition to use as an emulsifier, it is also an anti-caking and thickening agent.
Specifications

Specifications

31566-31-1
Poly Pail
12 kg
CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO
358.56
100%
C21H42O4
VBICKXHEKHSIBG-UHFFFAOYNA-N
2,3-dihydroxypropyl octadecanoate
Food Grade

Chemical Identifiers

31566-31-1
358.56
2,3-dihydroxypropyl octadecanoate
C21H42O4
VBICKXHEKHSIBG-UHFFFAOYNA-N
CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions