missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Monostearate, Powder, Food Grade, Spectrum™ Chemical
GL149, 31566-31-1, C21H42O4
Supplier: Spectrum Chemical Mfg Cor GL14925KG
Description
Spectrum™ Chemical Glyceryl Monostearate, Powder, Food Grade is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color nor odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. In addition to use as an emulsifier, it is also an anti-caking and thickening agent.Specifications
| 31566-31-1 | |
| Poly Pail | |
| 2.5 kg | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO | |
| 358.56 |
| 100% | |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| 2,3-dihydroxypropyl octadecanoate | |
| Food Grade |
Chemical Identifiers
| 31566-31-1 | |
| 358.56 | |
| 2,3-dihydroxypropyl octadecanoate |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO |