missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Monostearate, Flake, Spectrum™ Chemical
GL141, 31566-31-1, C21H42O4
$285.06 - $5645.27
Chemical Identifiers
| CAS | 31566-31-1 |
|---|---|
| Molecular Formula | C21H42O4 |
| Molecular Weight (g/mol) | 358.56 |
| InChI Key | VBICKXHEKHSIBG-UHFFFAOYNA-N |
| IUPAC Name | 2,3-dihydroxypropyl octadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602998
|
Spectrum Chemical Mfg Cor
GL141100GM |
100 g | Amber Glass Bottle |
Each for $285.06
|
|
||||
|
18602995
|
Spectrum Chemical Mfg Cor
GL141500GM |
500 g | Amber Glass Bottle |
Each for $327.82
|
|
||||
|
18602996
|
Spectrum Chemical Mfg Cor
GL14125KG |
2.5 kg | Poly Pail |
Each for $973.67
|
|
||||
|
18602999
|
Spectrum Chemical Mfg Cor
GL14145KGBL |
45 kg | Fiber Drum |
Each for $5,645.27
|
|
||||
Description
Spectrum™ Chemical Glyceryl Monostearate, Flake is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color or odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. It is used as a food additive and emulsifier, as well as an anti-caking and thickening agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 31566-31-1 | |
| 358.56 | |
| 2,3-dihydroxypropyl octadecanoate |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO |
Specifications
| 31566-31-1 | |
| Amber Glass Bottle | |
| 100 g | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO | |
| 358.56 |
| 100% | |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| 2,3-dihydroxypropyl octadecanoate | |
| Reagent |