missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Monostearate, Flake, Spectrum™ Chemical
GL141, 31566-31-1, C21H42O4
Supplier: Spectrum Chemical Mfg Cor GL141500GM
Description
Spectrum™ Chemical Glyceryl Monostearate, Flake is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color or odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. It is used as a food additive and emulsifier, as well as an anti-caking and thickening agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 31566-31-1 | |
| Amber Glass Bottle | |
| 500 g | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO | |
| 358.56 |
| 100% | |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| 2,3-dihydroxypropyl octadecanoate | |
| Reagent |
Chemical Identifiers
| 31566-31-1 | |
| 358.56 | |
| 2,3-dihydroxypropyl octadecanoate |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO |