missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monostearate, Flake, Spectrum™ Chemical

Catalog No. 18602999
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602999 45 kg Amber Glass Bottle
18602998 100 g Amber Glass Bottle
18602995 500 g Amber Glass Bottle
18602996 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602999 Supplier Spectrum Chemical Mfg Cor Supplier No. GL14145KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

GL141, 31566-31-1, C21H42O4

Spectrum™ Chemical Glyceryl Monostearate, Flake is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color or odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. It is used as a food additive and emulsifier, as well as an anti-caking and thickening agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 31566-31-1
Molecular Formula C21H42O4
Molecular Weight (g/mol) 358.56
InChI Key VBICKXHEKHSIBG-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxypropyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 45 kg
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material Glyceryl Monostearate, Flake