missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monooleate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013742
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013742 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013742 Supplier Merck Emd Millipore Supplier No. PHR14841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glyceryl Monooleate, commonly known as monoolein, is a glycerol monoester of oleic acid. Pharmaceutically, it is used as an emulsifier and permeation enhancer.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 25496-72-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H40O4
Synonym Glycerol mono-oleate
Type Certified Reference Material
Quantity 1 g
Formula Weight 356.54
Grade Certified Reference Material