missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, pure, 83.5-88.5 wt% aqueous solution
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
$190.26 - $190.26
Chemical Identifiers
| CAS | 56-81-5 |
|---|---|
| Molecular Formula | C3H8O3 |
| Molecular Weight (g/mol) | 92.09 |
| MDL Number | MFCD00004722 |
| InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
| Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
| PubChem CID | 753 |
| ChEBI | CHEBI:17754 |
| IUPAC Name | propane-1,2,3-triol |
| SMILES | OCC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC277360010
|
Thermo Scientific Chemicals
277360010 |
1 L | Glass bottle |
Each for $190.26
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Specifications
| 56-81-5 , 7732-18-5 | |
| Colorless | |
| 290.0°C | |
| Glass bottle | |
| 1.4520 to 1.4750 | |
| MFCD00004722 | |
| 01, 502 | |
| 15, 4520 | |
| Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform and in, fixed and volatile oils | |
| OCC(O)CO | |
| 92.09 | |
| 1069 mPa.s (20°C) | |
| 92.09 | |
| Pure | |
| Glycerol |
| 18.0°C | |
| 1.2270g/mL | |
| 160°C | |
| C3H8O3 | |
| HOCH2CH(OH)CH2OH | |
| 1 L | |
| 1.227 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| CHEBI:17754 | |
| 83.5 to 88.5% | |
| Viscous Liquid |
Safety and Handling
EINECSNumber : 200-289-5
RTECSNumber : MA8050000
TSCA : TSCA
RUO – Research Use Only