missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Molecular Biology Grade, Hoefer™

Numéro de catalogue. 03500507
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
03500507 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 03500507 Fournisseur Hoefer Inc Code fournisseur GR1241
Il en reste null
PAGE, density agent, sedimentation medium

Identifiants chimiques

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Spécifications

Color Colorless
Assay Percent Range ≥99.5%
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
Formula Weight 92.09
Grade Molecular Biology
Physical Form Viscous Liquid
Chemical Name or Material Glycerol