missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Glycerol, Calbiochem™,
Supplier: MilliporeSigma™ 3563501000ML
Specifications
| Glycerol | |
| Colorless | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| OCC(O)CO | |
| 92.09 | |
| CHEBI:17754 | |
| ≥99% | |
| Liquid |
| 56-81-5 | |
| C3H8O3 | |
| 1 L | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| 92.1 | |
| GC |
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Safety and Handling
RTECSNumber : MA8050000
Recommended Storage : 15° to 30°C (59° to 86°F)