missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycerol, Calbiochem™,

Catalog No. p-5584245
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
356350500ML 500 mL
3563501000M 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 356350500ML Supplier MilliporeSigma™ Supplier No. 356350500ML
Only null left

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Quantity 500 mL
Formula Weight 92.1
Percent Purity ≥99%
Grade GC
Physical Form Liquid
Chemical Name or Material Glycerol
RTECSNumber MA8050000
Recommended Storage 15° to 30°C (59° to 86°F)