missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002937
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002937 500 mL Narrow Mouth Amber Glass Bottle
02002935 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002937 Supplier Avantor Supplier No. 213601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Flash Point 199.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.26kg
Formula Weight 92.1
Percent Purity ≥99.5% (GC)
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Glycerol, Anhydrous