missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, >99%, Highly Purified, MP Biomedicals™
$83.24 - $191.18
Chemical Identifiers
| CAS | 56-81-5 |
|---|---|
| Molecular Formula | C3H8O3 |
| Molecular Weight (g/mol) | 92.09 |
| MDL Number | MFCD00004722 |
| InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
| Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
| PubChem CID | 753 |
| ChEBI | CHEBI:17754 |
| IUPAC Name | propane-1,2,3-triol |
| SMILES | OCC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19520480
|
MP Biomedicals Inc
0219520480 |
100 mL |
Each for $83.24
|
|
|||||
|
ICN19468090
|
MP Biomedicals Inc
0219468090 |
500 mL |
Each for $191.18
|
|
|||||
|
ICN19520491
|
MP Biomedicals Inc
0219520491 |
1 L |
Each for $179.25
|
|
|||||
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Specifications
| 56-81-5 | |
| 1.2613 g/cu cm at 20°C (Lit.) | |
| Odorless | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| OCC(O)CO | |
| 92.09 | |
| CHEBI:17754 | |
| Highly Purified | |
| Glycerol |
| Colorless | |
| >99% | |
| C3H8O3 | |
| 100 mL | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| 92.09 | |
| Syrupy Liquid |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C).