missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, >99%, Highly Purified, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219468090

 View more versions of this product

Catalog No. ICN19468090


Only null left

Description

Description

Specifications

Specifications

Glycerol
Colorless
>99%
C3H8O3
500 mL
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
Syrupy Liquid
56-81-5
1.2613 g/cu cm at 20°C (Lit.)
Odorless
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
OCC(O)CO
92.09
CHEBI:17754
Highly Purified

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15 to 30°C).

SDS
Documents

Documents

Promotions

Promotions