missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014658
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014658 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014658 Supplier Merck Emd Millipore Supplier No. PHR10205G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 56-81-5
Vapor Pressure <1 mmHg (20°C)
Vapor Density 3.1 (vs. air)
Density 1.25 g/mL
Physical Form Neat
Linear Formula HOCH2CH(OH)CH2OH
MDL Number MFCD00004722
Melting Point 20°C
Boiling Point 182°C (20 mmHg)
Synonym Glycerol; 1,2,3-Propanetriol; Glycerin
Quantity 5 g
Formula Weight 92.09
Grade Certified Reference Material
Show More Show Less