missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, Natural, EP, BP, USP, 99-101%, Spectrum™ Chemical

G1016, 56-81-5, C3H8O3

Supplier:  Spectrum Chemical Mfg Cor G101620LTPL

 View more versions of this product

Catalog No. 18602966

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Glycerin, Natural, USP, EP, BP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Synthetic, USP, EP, BP, JP . Natural glycerin is produced from the hydrolysis of animal or vegetable lipids during soap manufacture but the majority of natural glycerin now comes as a result of it being a side product during the production of biodiesel. All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

56-81-5
0.01%
C3H8O3
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
99 to 101%
100%
Poly Drum
1.470-1.475
20 L
OCC(O)CO
92.09
Multi Compendia

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
OCC(O)CO
C3H8O3
MFCD00004722
propane-1,2,3-triol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions