missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, Natural, EP, BP, USP, 99-101%, Spectrum™ Chemical

Catalog No. p-7157482
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602959 500 mL Amber Glass Bottle
18602958 500 mL Poly Bottle
18602968 1 L Poly Bottle
18602960 6 x 500 mL Amber Glass Bottle
18602961 6 x 500 mL Poly Bottle
18602962 4 L Amber Glass Bottle
18602963 4 L Poly Bottle
18602964 4 x 4 L Amber Glass Bottle
18602965 4 x 4 L Poly Bottle
18602966 20 L Poly Drum
18602967 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18602959 Supplier Spectrum Chemical Mfg Cor Supplier No. G1016500MLGL
Only null left

G1016, 56-81-5, C3H8O3

Spectrum™ Chemical Glycerin, Natural, USP, EP, BP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Synthetic, USP, EP, BP, JP . Natural glycerin is produced from the hydrolysis of animal or vegetable lipids during soap manufacture but the majority of natural glycerin now comes as a result of it being a side product during the production of biodiesel. All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.01%
Packaging Amber Glass Bottle
Refractive Index 1.470-1.475
Quantity 500 mL
Percent Purity 99 to 101%
Grade Multi Compendia