missing translation for 'onlineSavingsMsg'
Learn More

Glycerin (Glycerol), 50% (v/v), Ricca Chemical

Catalog No. 329016
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
329016 500 mL Natural Poly Bottle
329032 1 L Natural Poly Bottle
32905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 329016 Supplier Ricca Chemical Company Supplier No. 329016
Only null left

Glycerin (Glycerol), 50% (v/v) Aqueous Solution

Chemical Identifiers

CAS 56-81-5 , 7732-18-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Min % 55.0, 43.29
CAS Max % 57.0, 45.06
Percent Purity 50%
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material Glycerin