missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glutathione Sepharose™ 4B Media
Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. Cytiva Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.
$517.51 - $8711.30
Specifications
| Material | Glutathione Sepharose 4 B |
|---|---|
| pH Range | 4 to 13 |
| Particle Size | 90 μm |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
45000139
|
Cytiva
17075601 |
10 mL |
Each for $517.51
|
|
|||||
|
45-002-065
|
Cytiva
17075605 |
100 mL |
Each for $2,925.71
|
|
|||||
|
45-000-140
|
Cytiva
17075604 |
300 mL |
Each for $8,711.30
|
|
|||||
Description
- Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
- Recommended flowrate (H2O at room temp.): 75cm/hour
- Stable in presence of all commonly used aqueous buffers
- Stable in pH range 4 to 13
Specifications
| Glutathione Sepharose 4 B | |
| 4 to 13 | |
| 90 μm |