missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glutathione Sepharose™ 4B Media
Description
- Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
- Recommended flowrate (H2O at room temp.): 75cm/hour
- Stable in presence of all commonly used aqueous buffers
- Stable in pH range 4 to 13
Specifications
Specifications
| Material | Glutathione Sepharose 4 B |
| pH Range | 4 to 13 |
| Particle Size | 90 μm |
| Quantity | 100 mL |