missing translation for 'onlineSavingsMsg'
Learn More

Glutarimide 98.0+%, TCI America™

Catalog No. G02695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02695G Supplier TCI America Supplier No. G02695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1121-89-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00006670
InChI Key KNCYXPMJDCCGSJ-UHFFFAOYSA-N
Synonym glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci
PubChem CID 70726
ChEBI CHEBI:5435
IUPAC Name piperidine-2,6-dione
SMILES O=C1CCCC(=O)N1

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 113.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Glutarimide
RTECSNumber MA4000000
TSCA No