missing translation for 'onlineSavingsMsg'
Learn More

Glutarimide, 98%

Catalog No. AAL0096822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0096822 100 g
AAL0096806 5 g
AAL0096814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0096822 Supplier Thermo Scientific Chemicals Supplier No. L0096822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1121-89-7 | C5H7NO2 | 113.12 g/mol

Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1121-89-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00006670
InChI Key KNCYXPMJDCCGSJ-UHFFFAOYSA-N
Synonym glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci
PubChem CID 70726
ChEBI CHEBI:5435
IUPAC Name piperidine-2,6-dione
SMILES O=C1CCCC(=O)N1

Specifications

Melting Point 153°C to 157°C
Odor Nearly Odorless
Quantity 100 g
Beilstein 110052
Solubility Information Soluble in water,hot ethanol and boiling benzene. Insoluble in ether.
Formula Weight 113.12
Percent Purity 98%
Chemical Name or Material Glutarimide
EINECSNumber 214-340-4
RTECSNumber MA4000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only