missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaric Dialdehyde, 50% in Water, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor G3060500ML
Specifications
| 111-30-8,7732-18-5 | |
| Amber Glass Bottle | |
| 500 mL | |
| Reagent |
Chemical Identifiers
| 111-30-8 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.