missing translation for 'onlineSavingsMsg'
Learn More

Glutaric anhydride, 98%

Catalog No. AAA1115218
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1115218 50 g
AAA1115222 100 g
AAA1115230 250 g
AAA111520B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1115218 Supplier Thermo Scientific Chemicals Supplier No. A1115218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-55-4 | C5H6O3 | 114.1 g/mol

Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.

Chemical Identifiers

CAS 108-55-4
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006679
InChI Key VANNPISTIUFMLH-UHFFFAOYSA-N
Synonym glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
PubChem CID 7940
IUPAC Name oxane-2,6-dione
SMILES C1CC(=O)OC(=O)C1

Specifications

Melting Point 52°C to 57°C
Density 1.411 g/mL
Boiling Point 291°C
Flash Point 178°C (352°F)
Assay Percent Range 98%
Quantity 50 g
Beilstein 110051
Sensitivity Moisture sensitive
Solubility Information Soluble in Ether,dimethyl sulfoxide and methanol and water.
Formula Weight 114.1
Percent Purity 98%
Chemical Name or Material Glutaric anhydride
Show More Show Less
Hazard Category H302-H312-H315-H318
Hazard Statement GHS H Statement
H318-H302-H312-H335-H315
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 203-593-6
RTECSNumber MA3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only