missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose Standard, 10,000 ppm C6H12O6 in Aqueous Benzoic Acid Solution, Ricca Chemical
$61.72 - $101.19
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
| PubChem CID | 107526 |
| ChEBI | CHEBI:42758 |
| IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
| SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
3280-4
|
Ricca Chemical Company
32804 |
120 mL | Natural Poly Bottle |
Each for $61.72
|
|
||||
|
R3280000500
|
Ricca Chemical Company
R3280000500A |
500 mL | Natural Poly Bottle |
Each for $101.19
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| 107526 | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
| C6H12O6 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| CHEBI:42758 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
Specifications
| Natural Poly Bottle | |
| 97.0,0.43,0.18 | |
| C6H12O6 | |
| Standard | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| Mixture | |
| 107526 | |
| Laboratory |
| 7732-18-5,50-99-7,65-85-0 | |
| 100.0,0.45,0.19 | |
| 100°C | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
| 120 mL | |
| CHEBI:42758 | |
| Ricca Chemical Company Gluclose Standard |