missing translation for 'onlineSavingsMsg'
Learn More

Glucose Standard, 10,000 ppm C6H12O6 in Aqueous Benzoic Acid Solution, Ricca Chemical

Catalog No. 32804
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
32804 120 mL Natural Poly Bottle
R3280000500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32804 Supplier Ricca Chemical Company Supplier No. 32804
Only null left

Chemical Identifiers

CAS 7732-18-5 , 50-99-7 , 65-85-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) Mixture
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 0.43, 0.18
CAS Max % 100.0, 0.45, 0.19
Boiling Point 100°C
Type Standard
Quantity 120 mL
Grade Laboratory
Chemical Name or Material Ricca Chemical Company Gluclose Standard