missing translation for 'onlineSavingsMsg'
Learn More

Glucose-6-phosphate dehydrogenase

Catalog No. AAJ611814I
Change view
Click to view available options
Quantity:
1 kU
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ611814I 1 kU Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ611814I Supplier Thermo Scientific Chemicals Supplier No. J611814I

CAS: 9001-40-5

Glucose-6-phosphate dehydrogenase, yeast converts p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+ for the determination of blood glucose or creatine kinase. It is used to investigate the effect of dehydroepiandrosterone on cell growth and mitochondrial function in TM-3 cells, in bovine follicular cells and in the incidence of apoptosis and transcript abundance. It is also useful to test ketose reductase activity in developing maize endosperm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9001-40-5
MDL Number MFCD00081656
Synonym EC 1.1.1.49,RP-87086

Specifications

Packaging Plastic bottle
Quantity 1 kU
Solubility Information Soluble in water and dilute buffer.
Physical Form Lyophilized Powder
Chemical Name or Material Glucose-6-phosphate dehydrogenase
EINECSNumber 232-602-6
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only