missing translation for 'onlineSavingsMsg'
Learn More

Glucose, 20% (w/v), Ricca Chemical

Catalog No. R3254000100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R3254000100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3254000100 Supplier Ricca Chemical Company Supplier No. R3254000100A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) Mixture
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

CAS Min % 82.0, 16.34
CAS Max % 85.0, 17.01
Color Colorless
Packaging Natural Poly Bottle
Quantity 100 mL
Percent Purity 20% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 20% (w/v) Aqueous Solution
Physical Form Liquid
Chemical Name or Material Glucose
ShelfLife 12 months