missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gluconolactone, USP, 99-101%, Spectrum™ Chemical
G1165, 90-80-2, C6H10O6
$621.76 - $1834.39
Chemical Identifiers
| CAS | 90-80-2 |
|---|---|
| Molecular Formula | C6H10O6 |
| Molecular Weight (g/mol) | 178.14 |
| MDL Number | MFCD00006647 |
| InChI Key | PHOQVHQSTUBQQK-UHFFFAOYNA-N |
| IUPAC Name | 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one |
| SMILES | OCC1OC(=O)C(O)C(O)C1O |
Description
Spectrum™ Chemical Gluconolactone, USP is an exfoliant and used to treat acne lesions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 90-80-2 | |
| 178.14 | |
| PHOQVHQSTUBQQK-UHFFFAOYNA-N | |
| OCC1OC(=O)C(O)C(O)C1O |
| C6H10O6 | |
| MFCD00006647 | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one |
Specifications
| 90-80-2 | |
| Amber Glass Bottle | |
| MFCD00006647 | |
| PHOQVHQSTUBQQK-UHFFFAOYNA-N | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one | |
| 99 to 101% |
| 100% | |
| C6H10O6 | |
| 1 kg | |
| OCC1OC(=O)C(O)C(O)C1O | |
| 178.14 | |
| USP |