missing translation for 'onlineSavingsMsg'
Learn More

Gluconolactone, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18602929
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602929 1 kg Amber Glass Bottle
18602930 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602929 Supplier Spectrum Chemical Mfg Cor Supplier No. G11651KG

G1165, 90-80-2, C6H10O6

Spectrum™ Chemical Gluconolactone, USP is an exfoliant and used to treat acne lesions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 99 to 101%
Grade USP