accessibility menu, dialog, popup

UserName

HOOOOHHOHO

CAS RN 90-80-2

IUPAC Name: 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
Synonyms: gluconolactone
Molecular Weight (g/mol): 178.14
Molecular Formula: C6H10O6
InChi Key: PHOQVHQSTUBQQK-UHFFFAOYNA-N
SMILES: OCC1OC(=O)C(O)C(O)C1O
Percent Purity 
  • (2)
  • (4)
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (5)
  • (4)
Grade 
  • (4)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (6)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (2)
  • (2)