missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gluconic Acid (contains Gluconolactone) (45-50% in Water), TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Physical Form | Liquid |
| UN Number | 3265 |
| Formula Weight | 196.16 |
| Chemical Name or Material | Gluconic Acid (contains Gluconolactone) (45-50% in Water) |
Safety and Handling
| EINECSNumber | (2)-1409 |
| RTECSNumber | LZ5057100 |
| TSCA | Yes |