missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Innovatek Medical Inc. GlucoCheck Multi-Patient Blood Glucose Test Strips
Description
- For use with the GlucoCheck Multi-Patient Blood Glucose Meter
- Quantitative glucose measurement in fresh fingertip capillary whole blood
- 5-second results; meter measurement range: 1.1-33.3 mmol/L
- Single-use strips; no coding required
- Store at 1-30°C in the original vial; do not freeze
- Opened and unopened vials may be used until labeled expiry when stored and handled as directed
- 50 strips per vial; 2 vials per kit (100 strips)
Spécifications
Spécifications
| Marquage CE | No |
| Analytes détectables | Glucose |
| Catégorie | Test Strips |
| Forme | Solid |