missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Innovatek Medical Inc. GlucoCheck Blood Glucose Control Solution, A and B
Description
- For use with GlucoCheck Multi-Patient Blood Glucose Meter and Test Strips
- Checks system performance and test technique
- Bi-level controls A and B; 2 vials per kit, 4 mL each
- Contains D-glucose, preservative, and red dye
- Store tightly capped at 8-30°C; do not refrigerate or freeze
- Use before labeled expiry and within 3 months (90 days) of opening
- Bring controls, meter, and test strips to 20-25°C before testing
- Compare results with the corresponding control range printed on the test strip vial
Spécifications
Spécifications
| Marquage CE | No |
| Analytes détectables | Glucose |
| Catégorie | Control Solution |
| Forme | Liquid |