Learn More
Globe Scientific Test Tubes
Multi-use test tubes to fit most laboratory applications
 Afficher plus de versions de ce produit
Numéro de catalogue. 361016879
Il en reste null
Description
Description
  • Non-sterile
  • Molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents
  • Polystyrene tubes can withstand 3000rcf
  • Polypropylene tubes can withstand 5000rcf and are autoclavable and freezable
Spécifications
Identifiants chimiques
Test Tube
Spécifications
8 mL
Polypropylene
No
16 mm
75 mm
1000/Cs.
Autoclavable
Test Tube
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS