Learn More
Globe Scientific Sterile Rigid 10uL & 1uL Combo Inoculation Loops
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Supplier: Globe Scientific 2860F
Description
- Rigid, medical grade of polystyrene (PS) construction
- 10μL and 1μL
- Sterilized by gamma radiation
- White
- Available individually wrapped or in peel packs
Chemical Identifiers
| Polystyrene | |
| Single-use | |
| Needle and calibration certificate |
| Sterile | |
| Combo Inoculation Loop |
Specifications
| Single-use | |
| Sterile | |
| White | |
| Needle and calibration certificate | |
| Peel Pack |
| 10 μL | |
| Polystyrene | |
| Combo Inoculation Loop | |
| 10 |