missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Glipizide | |
| 206°C | |
| C21H27N5O4S | |
| 1 g | |
| ZJJXGWJIGJFDTL-UHFFFAOYSA-N | |
| N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide | |
| 3478 | |
| 445.54 | |
| Crystalline Powder |
| 29094-61-9 | |
| White-Yellow | |
| MFCD00072159 | |
| glipizide, glucotrol, glydiazinamide, melizide, glibenese, glucozide, glupizide, sucrazide, dipazide, glupitel | |
| CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3 | |
| 445.538 | |
| CHEBI:5384 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 29094-61-9 | |
| 445.538 | |
| ZJJXGWJIGJFDTL-UHFFFAOYSA-N | |
| 3478 | |
| N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide |
| C21H27N5O4S | |
| MFCD00072159 | |
| glipizide, glucotrol, glydiazinamide, melizide, glibenese, glucozide, glupizide, sucrazide, dipazide, glupitel | |
| CHEBI:5384 | |
| CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3 |
Safety and Handling
RTECSNumber : YS7640000
TSCA : No