missing translation for 'onlineSavingsMsg'
Learn More

Glipizide 98.0+%, TCI America™

Catalog No. p-7130251
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03691G 1 g
G03695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03691G Supplier TCI America Supplier No. G03691G

Chemical Identifiers

CAS 29094-61-9
Molecular Formula C21H27N5O4S
Molecular Weight (g/mol) 445.538
MDL Number MFCD00072159
InChI Key ZJJXGWJIGJFDTL-UHFFFAOYSA-N
Synonym glipizide, glucotrol, glydiazinamide, melizide, glibenese, glucozide, glupizide, sucrazide, dipazide, glupitel
PubChem CID 3478
ChEBI CHEBI:5384
IUPAC Name N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
SMILES CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 1 g
Formula Weight 445.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glipizide
RTECSNumber YS7640000
TSCA No