missing translation for 'onlineSavingsMsg'
Learn More

GlcNAc beta(1-3)GalNAc-alpha-pNP 98.0+%, TCI America™

Catalog No. G03765MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03765MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03765MG Supplier TCI America Supplier No. G03765MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125455-64-3
Molecular Formula C22H31N3O13
Molecular Weight (g/mol) 545.498
InChI Key HXQAUFSCNOLKJP-GVDFRNMQSA-N
PubChem CID 72943121
IUPAC Name N-[(3R,4R,5S,6R)-2-[(2R,3R,4R,5R,6R)-5-acetamido-3-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-4-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OC2C(C(OC(C2O)CO)OC3=CC=C(C=C3)[N+](=O)[O-])NC(=O)C)CO)O)O

Specifications

Color White
Quantity 5 mg
Formula Weight 545.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNAc beta(1-3)GalNAc-alpha-pNP
TSCA No
Recommended Storage Freezer