Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10
Catalog No. VT87146
Encompass
Change view
Click to view available options
Quantity:
100 Pack
50 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VT87146 100 Pack
22749754 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. VT87146 Supplier Kova Plastics by Alltrista Supplier No. 87146E
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Precision manufactured, 10-chambered slide available without a hemocytometer-type grid
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis.
  • Made from optically clear plastic
  • Glasstic Slide features 10 individual fully segregated specimen wells
  • Capillary action reduces the likelihood of pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
  • Correlate well with results obtained by using a hemocytometer: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Specifications
Type Slide 10
For Use With (Application) Microscopy
Gridded No
Material Plastic with the optical qualities of glass
No. of Wells 10
Quantity 100 Pack