missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Glass Tubing Assortment

Catalog No. S15780
Change view
Click to view available options
Diameter (Metric) Outer:
Various
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
S15780 Various
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15780 Supplier United Scientific™ Supplier No. GBT001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made from Borosilicate Glass

  • This convenient one-pound assortment of borosilicate glass tubing includes various thicknesses and wall sizes.
  • All pieces are 12 in. length

Specifications

Autoclavable Autoclavable
Diameter (English) Inner Various
Diameter (Metric) Inner Various
Material Borosilicate Glass (3.3)
Diameter (English) Outer Various
Diameter (Metric) Outer Various
Overall Length (English) 12 in.
Overall Length (Metric) 30.48 cm
For Use With Glass Cutter
Color Clear
Product Type Glass Tubing
Show More Show Less