missing translation for 'onlineSavingsMsg'
Learn More

Glass Spheres , MilliporeSigma™ Supelco™

Catalog No. 111015100
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015100 100 g
111015099 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015100 Supplier Merck Emd Millipore Supplier No. 440345100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silica is a naturally occurring material in minerals, flint and in some plants in crystalline phase. Silica used in industries is in synthetic form. Surface area, pore volume, pore size and particle size are independently controllable to some extent. The crystalline silica may be classified based on atmospheric pressure as:a. Quartz = 1143Kb. Tridymite 1143 - 1743Kc. Cristobalite = 1743, over 1973K it forms amorphous vitreous silica glass.

Specifications

CAS 65997-17-3
Density 1.1 g/mL (at 25°C)
Packaging Poly Bottle
Linear Formula O2Si
MDL Number MFCD00147032
Quantity 100 g
Formula Weight 60.08
Grade High-Purity Grade
Physical Form Powder