missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Rectangular Block

With all angles accurately worked and optically worked surfaces.

$38.75 - $50.29

Spécifications

Product Type Index of Refraction Plate
Lens Material Glass
For Use With (Application) Studying refraction and geometrical optics in physics and physical science classes
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W x H) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W x H) Prix Quantity & Availability  
S24378
Afficher les documents Détails de la promotion
Eisco™
PH0566C
100 x 60 x 18 mm
chaque for $38.75
Il en reste null
 
S24379
Afficher les documents Détails de la promotion
Eisco™
PH0566D
115 x 65 x 18 mm
chaque for $50.29
Il en reste null
 
Description

Description

  • Made from imported optical glass
  • Glass free from bubbles
Spécifications

Spécifications

Index of Refraction Plate
Glass
Studying refraction and geometrical optics in physics and physical science classes
Videos
FDS
Documents

Documents