missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Powder Funnels
Description
- Made from high quality borosilicate 3.3 glass
- Ideal for transferring powder reagents
- 60° angle stem
- Autoclavable
Warranty
1 year limited manufacturer warranty
Spécifications
Spécifications
| Diameter (Metric)Top | 120 mm |
| Material | Borosilicate Glass |
| Height (Metric) | 10.2 cm |
| Autoclavable | Autoclavable |
| Color | Clear |
| Graduated | No |
| For Use With (Application) | Transferring powder |
| Quantity | 1 |
| Length (English) Overall | 4.85 in. |
| Length (Metric) Overall | 12.31 cm |
| Afficher plus de résultats |