missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Powder Funnels

Numéro de catalogue. S89486
Change view
Click to view available options
Diameter (Metric)Top:
45 mm
55 mm
75 mm
85 mm
100 mm
120 mm
Height (Metric):
7.6 cm
10.2 cm
12.7 cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Diameter (Metric)Top Height (Metric)
S89486 100 mm 10.2 cm
S89482 45 mm 12.7 cm
S89483 55 mm 12.7 cm
S89484 75 mm 12.7 cm
S89485 85 mm 7.6 cm
S89487 120 mm 10.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. S89486 Fournisseur Eisco™ Code fournisseur CH0459E
Il en reste null

Eisco™ Glass Powder Funnels designed with 60° angle stems are ideal for transferring powder reagents.

  • Made from high quality borosilicate 3.3 glass
  • Ideal for transferring powder reagents
  • 60° angle stem
  • Autoclavable

Warranty

1 year limited manufacturer warranty

Spécifications

Diameter (Metric)Top 100 mm
Material Borosilicate Glass
Height (Metric) 10.2 cm
Autoclavable Autoclavable
Color Clear
Graduated No
For Use With (Application) Transferring powder
Quantity 1
Height (English) 4 in.
Product Type Filling Funnel