missing translation for 'onlineSavingsMsg'
Learn More

Girard's Reagent P 95.0+%, TCI America™

Catalog No. G003025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G003025G Supplier TCI America Supplier No. G003025G

Chemical Identifiers

CAS 1126-58-5
Molecular Formula C7H10ClN3O
Molecular Weight (g/mol) 187.627
MDL Number MFCD00011992
InChI Key NDXLVXDHVHWYFR-UHFFFAOYSA-N
Synonym 1-(Hydrazinocarbonylmethyl)pyridinium Chloride, (1-Pyridinio)acetohydrazide Chloride
PubChem CID 70773
IUPAC Name 2-pyridin-1-ium-1-ylacetohydrazide;chloride
SMILES C1=CC=[N+](C=C1)CC(=O)NN.[Cl-]

Specifications

Physical Form Powder/Crystals
Melting Point 203°C
Quantity 25 g
Formula Weight 187.63
Percent Purity ≥95.0% (T)
Chemical Name or Material Girard′s Reagent P