missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolide A, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013217
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013217 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013217 Supplier Merck Emd Millipore Supplier No. 5186310MGF
Only null left
Featured Industries: Food and Beverages

Specifications

CAS 15291-75-5
Physical Form Neat
Linear Formula C20H24O9
MDL Number MFCD00133365
Type Analytical Standard
Quantity 10 mg
Formula Weight 408.40
Percent Purity ≥98% (HPLC)
Grade Analytical Standard