missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain, Reagent Grade, LabChem™

Catalog No. LC148357
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC148357 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC148357 Supplier LabChem Supplier No. LC148357

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
Synonym azure a, dimethylthionine, giemsa stain, azur a, n,n-dimethylthionine, 3-amino-7-dimethylamino phenothiazin-5-ium chloride, methylene azure a, giemsa solution, 3-amino-7-dimethylaminophenazathionium chloride, unii-m731v243ef
PubChem CID 13735
IUPAC Name 7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Physical Form Powder
Melting Point 300°C
CAS Max % 100
Packaging Amber Glass
Solubility Soluble in water
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Color Golden/Green
Quantity 10 g
Linear Formula C14H14ClN3S
Formula Weight 291.8
Grade Reagent
Chemical Name or Material Giemsa Stain
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 257-438-2
Recommended Storage Room Temperature