missing translation for 'onlineSavingsMsg'
Learn More

Giemsa's azur-eosin-methylene blue, For Microscopy, MilliporeSigma™

Catalog No. M1092030100
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1092030100 100 g
M1092030025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1092030100 Supplier Technidata Supplier No. 1.09203.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For microscopy

Specifications

Chemical Name or Material Giemsa's azur-eosin-methylene blue
CAS 17372-87-1, 61-73-4
Odor Weak characteristic odor
Packaging Glass bottle
Color Dark Green
Quantity 100 g
Physical Form Solid