missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rabbit Serum

Standard (sterile-filtered)
$118.00 - $433.00
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
|---|---|
| Species | Rabbit |
| Age | 6 months or less |
| Country of Origin | United States |
| Form | Liquid |
Description
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Sera Category: Specialty
- Origin: United States
- Age: 6 months or less
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
| Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
| 6 months or less | |
| Liquid | |
| Standard (Sterile-filtered) | |
| Gibco |
| Rabbit | |
| United States | |
| Rabbit Serum | |
| Sterile | |
| Frozen |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.